Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondroitin sulfate A sodium salt, 90%
Description
Chondroitin sulfate is a sulfated (in different position) glycosaminoglycan (GAG).Composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. In chondroitin sulfate A, sulfate is on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar.
General Description
- Chondroitin sulfate A is characterized by a N-acetylgalactosamine molecule with a sulfated carbon-4
- Chondroitin sulfate A is found in the extracellular matrix and is an important structural polysaccharide
- Chondroitin sulfates play roles in cellular development and the formation of tissue barriers throughout the body
Application
- Chondroitin sulfate A is useful for in vitro study of nutrition and cellular-level repair
- Chondroitin sulfate A can be used in the laboratory study of cell signaling, tissue growth, and repair
Specifications
Specifications
| Chemical Name or Material | Chondroitin sulfate A sodium salt |
| Color | White |
| Quantity | 5 g |
| MDL Number | MFCD00130792 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 90% |
| Physical Form | Powder |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?