Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chondroitin sulfate (from pig) | 9007-28-7 | 95.1% | 5 G
SDP

Catalog No. 5000412378
Encompass_Preferred

Chondroitin sulfate from pig is a biochemical reagent suitable as a biological material or organic compound for life science research. This product is for research use only and not for patient use. It appears as an off-white to light yellow solid with a purity of 95.06%.

  • Suitable for life science related research
  • Can be used as a biological material or organic compound
  • Off-white to light yellow solid appearance
  • Purity of 95.06%
  • Formula: (C14H21NO14S)n
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Solubility in H2O is 100 mg/mL (requires ultrasonic)

Catalog No. 50-004-12378 Supplier Medchemexpress LLC Supplier No. HYB2162C5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chondroitin sulfate from pig is a biochemical reagent suitable as a biological material or organic compound for life science research. This product is for research use only and not for patient use. It appears as an off-white to light yellow solid with a purity of 95.06%.

  • Suitable for life science related research
  • Can be used as a biological material or organic compound
  • Off-white to light yellow solid appearance
  • Purity of 95.06%
  • Formula: (C14H21NO14S)n
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Solubility in H2O is 100 mg/mL (requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.