Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chondroitin sulfate sodium (from bovine) | 9007-28-7 | 99.4% | 10 G
SDP

Catalog No. 5000410468
Encompass_Preferred

Chondroitin sulfate from bovine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Formula: (C14H21NO14SNa)n
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility: 40 mg/mL in water (requires ultrasonic).

Catalog No. 50-004-10468 Supplier Medchemexpress LLC Supplier No. HYB2162B10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chondroitin sulfate from bovine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Formula: (C14H21NO14SNa)n
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility: 40 mg/mL in water (requires ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.