Learn More
Medisca Chondroitin Sulfate Sodium, USP (Bovine) (Dietary Supplement Grade)
Shop All Medisca Products

Description
Chondroitin Sulfate Sodium, USP (Bovine) comes as a white to off-white powder that is extremely hygroscopic once dried. It is freely soluble in water.
Alternative name(s): Bovine sodium chondroitin sulfate, Bovine sodium chondroitin sulphate,
Specifications
Specifications
| Chemical Name or Material | Chondroitin Sulfate Sodium, USP (Bovine) (Dietary Supplement Grade) |
| Packaging | 120cc Amber Glass Round w/m bottle |
| CAS | 9082-07-9 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | Not applicable |
| Quantity | 100 g |
| Percent Purity | ≥85.0 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.