Learn More
Sigma Aldrich Fine Chemicals Biosciences Chondroitinase ABC from Pr
Catalog No. 502905830
Encompass
Chondroitinase ABC is a lyase with specificity toward chondroitin sulfates and dermatan sulfates - with some minor activity toward hyaluronan. This recombinant version only contains the endolytic chondroitinase meaning it will not digest the non-reducing ends of proteoglycan substrates. It targets beta-(1 4)-linkages between N-acetylgalactosamine and either D-glucuronic acid or L-iduronic acid. The product of digestion is a mixture of Delta 4-unsaturated tetra- and di-saccharides.
Catalog No. 50-290-5830 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE01500.1ML
Chondroitinase ABC is a lyase with specificity toward chondroitin sulfates and dermatan sulfates - with some minor activity toward hyaluronan. This recombinant version only contains the endolytic chondroitinase meaning it will not digest the non-reducing ends of proteoglycan substrates. It targets beta-(1 4)-linkages between N-acetylgalactosamine and either D-glucuronic acid or L-iduronic acid. The product of digestion is a mixture of Delta 4-unsaturated tetra- and di-saccharides.