Learn More
Cayman Chemical Chonglou Saponin VII, Purity: ≥98%, CAS: 68124-04-9, 10 mg
SDP
Catalog No. 502919105
Encompass_Preferred
A steroid saponin with anticancer activity; inhibits the proliferation of HepG2 and HepG2/ADR cells at 1.98 and 2.97 µM; inhibits the proliferation of MDA-MB-231, MDA-MB-436, and MCF-7 breast cancer cells (IC50s = 3.16, 3.45, and 2.86 µM, respectively); induces intratumoral autophagy and reduces tumor growth in an MDA-MB-231 mouse xenograft model at 1.5 mg/kg
Catalog No. 50-291-9105 Supplier Cayman Chemical Supplier No. 3925110 MG
A steroid saponin with anticancer activity; inhibits the proliferation of HepG2 and HepG2/ADR cells at 1.98 and 2.97 µM; inhibits the proliferation of MDA-MB-231, MDA-MB-436, and MCF-7 breast cancer cells (IC50s = 3.16, 3.45, and 2.86 µM, respectively); induces intratumoral autophagy and reduces tumor growth in an MDA-MB-231 mouse xenograft model at 1.5 mg/kg