Learn More
Enzo Life Sciences CHOP monoclonal antibody (9C8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804551C100
Encompass_Preferred
Clone: 9C8. Isotype: IgG2b. Immunogen: Recombinant mouse CHOP (C/EBP homology protein). Species reactivity: Human, Mouse. Specificity: Recognizes an N-terminal epitope. Applications: ICC, IP, WB. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-9942