Learn More
AMPLYUS LLC miniPCR bio Chopped! Using CRISPR/Cas9 to cut DNA
SDP
Catalog No. NC2121969
Encompass
Chopped! Using CRISPR/Cas to Cut DNA: use authentic CRISPR/Cas in a test tube with a simple gel electrophoresis readout.
This item is not returnable. View return policy
Catalog No. NC2121969 Supplier AMPLYUS LLC Supplier No. KT180101
Chopped! Using CRISPR/Cas to Cut DNA: use authentic CRISPR/Cas in a test tube with a simple gel electrophoresis readout.