Learn More
Medchemexpress LLC Chordin-like 1/CHRDL 20ug | 20ug
SDP
Catalog No. 5000187391
Encompass_Preferred
The CHRDL1 protein acts as a BMP4 antagonist blocking its receptor interaction and redirecting neural stem cells from gliogenesis to neurogenesis The role of CHRDL1 in preventing glial cell fate adoption supports neuronal differentiation in the brain Chordin-like 1/CHRDL1 Protein Human (His solution) is the recombinant human-derived Chordin-like 1/CHRDL1 protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-87391 Supplier Medchemexpress LLC Supplier No. HYP7789620UG
The CHRDL1 protein acts as a BMP4 antagonist blocking its receptor interaction and redirecting neural stem cells from gliogenesis to neurogenesis The role of CHRDL1 in preventing glial cell fate adoption supports neuronal differentiation in the brain Chordin-like 1/CHRDL1 Protein Human (His solution) is the recombinant human-derived Chordin-like 1/CHRDL1 protein expressed by E coli with N-His labeled tag