Learn More
Sigma Aldrich Chorismic acid from Enterobacter aerogenes
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Intermediate in the biosynthesis of aromatic amino acids via the shikimate pathway.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
- C1761.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
- To learn more, visit sigma.com/metlin
Specifications
Specifications
| CAS | 617-12-9 |
| Molecular Formula | C10H10O6 |
| Linear Formula | C10H10O6 |
| MDL Number | MFCD00135935 |
| Quantity | 5 mg |
| Synonym | trans-3-([1-Carboxyethenyl]oxy)-4-hydroxy-1,5-cyclohexadiene-1-carboxylic acid |
| Molecular Weight (g/mol) | 226.18 |
| Percent Purity | ≥80% |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.