Learn More
TARGETMOL CHEMICALS INC CHPG HYDROCHLORIDE 5MG
SDP
Catalog No. 502581657
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHPG hydrochloride is a selective agonist of mGluR5.
Catalog No. 50-258-1657 Supplier TARGETMOL CHEMICALS INC Supplier No. T10809L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHPG hydrochloride is a selective agonist of mGluR5.