Learn More
Medchemexpress LLC Chpg sodium salt | 1303993-73-8 | 99.3% | 223.59 g/mol | C8H7ClNNaO3 | 1 ML
SDP
Catalog No. 5000211430
Encompass_Preferred
CHPG sodium salt is a selective metabotropic glutamate receptor 5 (mGluR5) agonist provided as a 10 mM solution in DMSO for research use. It has been used to study ERK and Akt signaling, oxidative stress, and neuroinflammation in cellular and in vivo models.
  • Selective mGluR5 agonist useful for receptor activation studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Characterized chemical identity with CAS 1303993-73-8.
  • Recommended storage at -80°C for long-term or -20°C for short-term, sealed and away from moisture.
Catalog No. 50-002-11430 Supplier Medchemexpress LLC Supplier No. HY101364A10MM 1ML
CHPG sodium salt is a selective metabotropic glutamate receptor 5 (mGluR5) agonist provided as a 10 mM solution in DMSO for research use. It has been used to study ERK and Akt signaling, oxidative stress, and neuroinflammation in cellular and in vivo models.
  • Selective mGluR5 agonist useful for receptor activation studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Characterized chemical identity with CAS 1303993-73-8.
  • Recommended storage at -80°C for long-term or -20°C for short-term, sealed and away from moisture.