Learn More
Medchemexpress LLC HY-15217 5mg , CHR-6494 CAS:1333377-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY152175MG
Encompass_Preferred
HY-15217 5mg CHR-6494 CAS: 1333377-65-3 CHR-6494 is a potent inhibitor of haspin, with an IC50 of 2 nM. CHR-6494 inhibits histone H3T3 phosphorylation. CHR-6494 can be used in the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0150