Learn More
MSC Sloan Vacuum Breaker, 1.5x9 in, Chrome Plated, 1 ea.
Catalog No. 502817959
Encompass
This vacuum breaker is a plumbing safety device designed to prevent back-siphonage of non-potable water into the potable water system, ensuring water quality and safety. Its chrome-plated finish offers enhanced durability and a polished aesthetic.
  • Prevents back-siphonage
  • Chrome plated
  • Dimensions: 1.5 in × 9 in
Catalog No. 50-281-7959 Supplier MSC Supplier No. 21006200
This vacuum breaker is a plumbing safety device designed to prevent back-siphonage of non-potable water into the potable water system, ensuring water quality and safety. Its chrome-plated finish offers enhanced durability and a polished aesthetic.
  • Prevents back-siphonage
  • Chrome plated
  • Dimensions: 1.5 in × 9 in