Learn More
Aviva Systems Biology Corp CHRAC1 Antibody - middle region (ARP39281_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39281_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRAC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHRAC1. Alternate names: CHARC1, CHARC15, CHRAC15, YCL1, CHRAC-1, CHRAC-15ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-56970