Learn More
Med Vet International Christmas Tree Connector, J0595C, 2 Pack
Catalog No. 5000581025
Encompass_Preferred
Dual fitting polypropylene. One end is a female DISS connector the other a barbed connector. One suggested use is from oxygen regulator connecting oxygen tubing to rescusitator bag. 2/pkg.
Catalog No. 50-005-81025 Supplier Med Vet International Supplier No. JOR595C
Dual fitting polypropylene. One end is a female DISS connector the other a barbed connector. One suggested use is from oxygen regulator connecting oxygen tubing to rescusitator bag. 2/pkg.