Learn More
Eurofins Discoverx ChemiScreen M4 Muscarinic Acetylcholine Receptor Membrane Preparation, 1 vial. (HTS117M)
Catalog No. 502005916
Encompass
The ChemiScreen M4 Muscarinic Acetylcholine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CHRM4 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5916 Supplier Eurofins Discoverx Supplier No. HTS117M
Add to Cart
Add to Cart
The ChemiScreen M4 Muscarinic Acetylcholine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CHRM4 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.