Learn More
Macherey-Nagel SPE columns CHROMABOND XTR, 150 mL, 37.5 g
Catalog No. NC1371860
Encompass_Preferred
CHROMABOND columns XTR - volume: 150 mL, content of sorbent: 37.5 g kieselguhr - for volumes up to 50 mL - material: PP, with PE filter elements - pack of 10
Catalog No. NC1371860 Supplier Macherey-Nagel Supplier No. 730509
CHROMABOND columns XTR - volume: 150 mL, content of sorbent: 37.5 g kieselguhr - for volumes up to 50 mL - material: PP, with PE filter elements - pack of 10