Learn More
Macherey-Nagel SPE columns CHROMABOND XTR, 6 mL, 1000 mg, BIG
Catalog No. NC0382342
Encompass_Preferred
CHROMABOND columns XTR BIGpack - Volume: 6 mL, contect of sorbent: 1000 mg kieselguhr - material: PP, with PE filter elements - packed in 5 bags with 50 pcs. - pack of 250
Catalog No. NC0382342 Supplier Macherey-Nagel Supplier No. 730487.250
CHROMABOND columns XTR BIGpack - Volume: 6 mL, contect of sorbent: 1000 mg kieselguhr - material: PP, with PE filter elements - packed in 5 bags with 50 pcs. - pack of 250