Learn More
Aviva Systems Biology Corp CHRNA1 Antibody - C-terminal region (AVARP13084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CHRNA1. Alternate names: ACHRA, ACHRD, CHRNA, CMS2A, FCCMS, SCCMSApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-53793