Learn More
Aviva Systems Biology Corp CHRNA1 Antibody - N-terminal region (ARP34909_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34909_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNA1. Alternate names: ACHRA, ACHRD, CHRNA, CMS2A, FCCMS, SCCMSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52980