Learn More
Aviva Systems Biology Corp CHRNA2 Antibody - N-terminal region (AVARP13064_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13064_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNA2. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53792