Learn More
Aviva Systems Biology Corp CHRNG Antibody - N-terminal region (AVARP13024_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13024_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNG. Alternate names: ACHRGApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62612