Learn More
Aviva Systems Biology Corp CHRNA3 Antibody - N-terminal region (ARP34964_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNA3. Alternate names: MGC104879, LNCR2, PAOD2, NACHRA3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55618