Learn More
Aviva Systems Biology Corp CHRNA5 Antibody - middle region (ARP34967_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34967_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHRNA5. Alternate names: LNCR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55620