Learn More
Aviva Systems Biology Corp CHRNA7 Antibody - N-terminal region (AVARP13018_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13018_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNA7. Alternate names: NACHRA7, CHRNA7-2ApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-53778