Learn More
Aviva Systems Biology Corp CHRNA9 Antibody - N-terminal region (AVARP13019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNA9. Alternate names: NACHRA9, HSA243342ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-53779