Learn More
Aviva Systems Biology Corp CHRNB2 Antibody - middle region (ARP37669_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37669_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHRNB2. Alternate names: EFNL3, nAChRB2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-56389