Learn More
Aviva Systems Biology Corp CHRNB2 Antibody - N-terminal region (AVARP13020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNB2. Alternate names: EFNL3, nAChRB2ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Zebrafish
Catalog No. 50-014-62605