Learn More
Aviva Systems Biology Corp CHRND Antibody - N-terminal region (ARP34975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRND. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CHRND. Alternate names: ACHRD, CMS2A, FCCMS, SCCMSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-438