Learn More
Aviva Systems Biology Corp CHRNE Antibody - N-terminal region (AVARP13023_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13023_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRNE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHRNE. Alternate names: ACHRE, CMS1D, CMS1E, CMS2A, FCCMS, SCCMSApplicationWB. Species reactivity: Guinea Pig, Mouse, Pig, Rat
Catalog No. 50-014-62610