Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 250mL Reservoir, 1in ID X 12in E.L., 2mm Stpk
Supplier: Chemglass Life Sciences CG120324
Column, Chromatography, 35/20 Socket Joint, 250mL Reservoir, 1in ID X 12in E.L., 2mm Stopcock. Similar to CG-1202 but with a COARSE porosity fritted disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with CG-1201 flow controller. Connection between the column and flow control adapter are spherical joints connected with CG-150 stainless steel clamps.