Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 500mL Reservoir, 1 1/2in ID X 18in E.L., 2mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG120328
Encompass_Preferred
Column, Chromatography, 35/20 Socket Joint, 500mL Reservoir, 1 1/2in ID X 18in E.L., 2mm Stopcock. Similar to CG-1202 but with a COARSE porosity fritted disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with CG-1201 flow controller. Connection between the column and flow control adapter are spherical joints connected with CG-150 stainless steel clamps.
Catalog No. CG120328