Learn More
Chemglass Life Sciences Column, Chromatography, 24/40 Outer Joint, 1/2in ID X 12in E.L., 2mm Stpk
SDP
Catalog No. CG118803
Encompass_Preferred
Column, Chromatography, 24/40 Outer Joint, 1/2in ID X 12in E.L., 2mm Stopcock General purpose column has a PTFE stopcock and a top outer joint. Glass wool (not supplied) is used to support column packing. Column is constructed using MEDIUM WALL TUBING. Can be used as a gravity eluted column or a flash column (as described by W.C. Still, M. Kahn and A. Mitra of Columbia University in JOURNAL OF ORGANIC CHEMISTRY 43, 2923 (1978), and covered in U.S. Patent Number 4,293,422 dated October 6, 1981) when used with optional CG-1191 flow controller and CG-1190 reservoir.
Catalog No. CG118803 Supplier Chemglass Life Sciences Supplier No. CG118803
Column, Chromatography, 24/40 Outer Joint, 1/2in ID X 12in E.L., 2mm Stopcock General purpose column has a PTFE stopcock and a top outer joint. Glass wool (not supplied) is used to support column packing. Column is constructed using MEDIUM WALL TUBING. Can be used as a gravity eluted column or a flash column (as described by W.C. Still, M. Kahn and A. Mitra of Columbia University in JOURNAL OF ORGANIC CHEMISTRY 43, 2923 (1978), and covered in U.S. Patent Number 4,293,422 dated October 6, 1981) when used with optional CG-1191 flow controller and CG-1190 reservoir.