Learn More
Chemglass Life Sciences Column, Chromatography, 3/4in ID X 10in E.L., 2mm Stpk
SDP
Catalog No. CG118706
Encompass_Preferred
Column, Chromatography, 3/4in ID X 10in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. CG118706 Supplier Chemglass Life Sciences Supplier No. CG118706
Column, Chromatography, 3/4in ID X 10in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.