Learn More
Chemglass Life Sciences Column, Chromatography, 3/4in ID X 12in E.L., 2mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG118607
Encompass_Preferred
Column, Chromatography, 3/4in ID X 12in E.L., 2mm Stopcock. General purpose gravity eluted column having a PTFE stopcock and reinforced beaded top. Column is constructed using MEDIUM WALL TUBING. Glass wool (not supplied) is used to support column packing. Lower end of the column has a round bottom.
Catalog No. CG118607