Learn More
Chemglass Life Sciences Column, Chromatography, 1in ID X 12in E.L., 2mm Stpk
SDP
Catalog No. CG118611
Encompass_Preferred
Column, Chromatography, 1in ID X 12in E.L., 2mm Stopcock. General purpose gravity eluted column having a PTFE stopcock and reinforced beaded top. Column is constructed using MEDIUM WALL TUBING. Glass wool (not supplied) is used to support column packing. Lower end of the column has a round bottom.
Catalog No. CG118611 Supplier Chemglass Life Sciences Supplier No. CG118611
Column, Chromatography, 1in ID X 12in E.L., 2mm Stopcock. General purpose gravity eluted column having a PTFE stopcock and reinforced beaded top. Column is constructed using MEDIUM WALL TUBING. Glass wool (not supplied) is used to support column packing. Lower end of the column has a round bottom.