Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 2000mL Reservoir, 3in ID X 8in E.L., 4mm Stpk
Supplier: Chemglass Life Sciences CG120335
Column, Chromatography, 35/20 Socket Joint, 2000mL Reservoir, 3in ID X 8in E.L., 4mm Stopcock. Similar to CG-1202 but with a COARSE porosity fritted disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with CG-1201 flow controller. Connection between the column and flow control adapter are spherical joints connected with CG-150 stainless steel clamps.