Learn More
Chemglass Life Sciences Column, Chromatography, 24/40 Outer Joint, 2in ID X 12in E.L., 2mm Stpk
SDP
Catalog No. CG118819
Encompass_Preferred
Column, Chromatography, 45/50 Outer Joint, 3in ID X 24in E.L., 4mm Stpk. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. CG118819 Supplier Chemglass Life Sciences Supplier No. CG118819
Column, Chromatography, 45/50 Outer Joint, 3in ID X 24in E.L., 4mm Stpk. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.