Learn More
Chemglass Life Sciences Column, Chromatography, 3in ID X 12in E.L., 4mm Stpk
SDP
Catalog No. CG118729
Encompass_Preferred
Column, Chromatography, 3in ID X 12in E.L., 4mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. CG118729 Supplier Chemglass Life Sciences Supplier No. CG118729
Column, Chromatography, 3in ID X 12in E.L., 4mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.