Learn More
Chemglass Life Sciences 500mL Reservoir, Chromatography, 24/40 Lower Inner
SDP
Catalog No. CG119012
Encompass_Preferred
500mL Reservoir, Chromatography, 24/40 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.
Catalog No. CG119012 Supplier Chemglass Life Sciences Supplier No. CG119012
500mL Reservoir, Chromatography, 24/40 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.