Learn More
Chemglass Life Sciences 2000mL Reservoir, Chromatography, 45/50 Lower Inner
SDP
Catalog No. CG119016
Encompass_Preferred
2000mL Reservoir, Chromatography, 45/50 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.
Catalog No. CG119016 Supplier Chemglass Life Sciences Supplier No. CG119016
2000mL Reservoir, Chromatography, 45/50 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.