Learn More
Chemglass Life Sciences Adapter, Flow Control, Chromatography, 45/50 Inner Rodaviss Joint
SDP
Catalog No. CG119511
Encompass_Preferred
Adapter, Flow Control, Chromatography, 45/50 Inner Rodaviss Joint. Flow control adapter for use with CG-1192 and CG-1193 chromatography columns along with CG-1194 reservoirs. Adapter has a 0-4mm Chem-Cap metering plug.
Catalog No. CG119511 Supplier Chemglass Life Sciences Supplier No. CG119511
Adapter, Flow Control, Chromatography, 45/50 Inner Rodaviss Joint. Flow control adapter for use with CG-1192 and CG-1193 chromatography columns along with CG-1194 reservoirs. Adapter has a 0-4mm Chem-Cap metering plug.