Learn More
Microsolv Technology Corporation Chrom Syringe 10Ml Ll Ns 100/P
Catalog No. 5001452801
Encompass_Preferred
Syringe, Chrom. Disposable, 10ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 100/PK
Catalog No. 50-014-52801 Supplier Microsolv Technology Corporation Supplier No. 58910SC
Syringe, Chrom. Disposable, 10ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 100/PK