Learn More
Microsolv Technology Corporation Chrom Syringe 10Ml Ll Ns 1000/
Catalog No. 5001452802
Encompass_Preferred
Syringe, Chrom. Disposable, 10ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 1000/CS
Catalog No. 50-014-52802 Supplier Microsolv Technology Corporation Supplier No. 58910SK
Syringe, Chrom. Disposable, 10ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 1000/CS