Learn More
Microsolv Technology Corporation Chrom Syringe 5Ml Ll Ns 100/Pk
Catalog No. 5001452798
Encompass_Preferred
Syringe, Chrom. Disposable, 5ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 100/PK.
Catalog No. 50-014-52798 Supplier Microsolv Technology Corporation Supplier No. 58905SC
Add to Cart
Add to Cart
Syringe, Chrom. Disposable, 5ml, Luer Lock, Non Sterile. MicroSolv Brand. Bulk Packed. 100/PK.