Learn More
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Shop All Thermo Scientific ProductsDescription
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.
For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.
- 8mm crimp top seal minimizes exposure between sample solvent and septum
- Neutral Borosilicate GOLD-Grade glass is a low expansion, high purity glass with an extremely low concentration of active sites to protect your analytes from unwanted interactions
- Precision-machined (glass or PTFE) sleeves allow the vials to be used in the vast majority of commercial autosamplers
Thermo Scientific™
Agilent™
Beckman™
Carlo Erba™
CTC™
Fisons™
PerkinElmer™
Shimadzu™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility
Specifications
Specifications
| Description | 0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass |
| Color | Clear |
| Diameter (Metric) Outer | 6 mm |
| Material | Glass |
| Volume (Metric) | 125 μL |
| Volume (Metric) Residual | <1 μL |
| Product Line | Chromacol Gold |
| Quantity | 500 |
| Closure Size | 8 mm |
| Marking Spot | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.