Learn More
FUJIFILM Biosciences Chromafenozide Standard
Catalog No. 5000450433
Encompass_Preferred
Chromafenozide Standard is a high-purity reference material designed for pesticide residue analysis with a minimum HPLC purity of 99 0% It is shipped and stored in glass containers requiring cool dark conditions 2 10 C to maintain stability Suitable for laboratories performing pesticide residue testing
Catalog No. 50-004-50433 Supplier FUJIFILM Biosciences Supplier No. 03318141
Chromafenozide Standard is a high-purity reference material designed for pesticide residue analysis with a minimum HPLC purity of 99 0% It is shipped and stored in glass containers requiring cool dark conditions 2 10 C to maintain stability Suitable for laboratories performing pesticide residue testing