Learn More
Macherey-Nagel Syringe filters CHROMAFIL Xtra PA-45/25
Supplier:  Macherey-Nagel 729213
Encompass_Preferred
CHROMAFIL Xtra disposable syringe filters PA-45/25 - membrane material: polyamide (Nylon) - pore dia.: 0.45 µm, membrane dia.: 25 mm - colorless PP case, labeled - pack of 100
Catalog No. NC1421917