Learn More
Macherey-Nagel Syringe filters CHROMAFIL O-45/15 MS
Supplier:  Macherey-Nagel 729009
Encompass_Preferred
CHROMAFIL disposable syringe filters O-45/15 MS - membrane material: polytetrafluoroethylene (PTFE) - pore dia.: 0.45 µm, membrane dia.: 15 mm - color code above: colorless / below: colorless - pack of 100
Catalog No. NC1208098